• USA Home
  • 522600 - Polycarbomethylsilane

EMAIL THIS PAGE TO A FRIEND
522600 Aldrich

Polycarbomethylsilane

average Mw ~2,000, electronic grade

Synonym: PCMS, Poly[(methylsilylene)methylene], Polycarbosilane

Purchase

Properties

Related Categories Materials Science, Other Silicone Polymers, Polymer Science, Polymers, Silicones More...
grade   electronic grade
mol wt   average Mw ~2,000
mp   >300 °C
density   1.1 g/mL at 25 °C(lit.)

Description

Application

Polymeric precursor of SiC ceramics. Ceramic coating from polymer melt or solution.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?