• USA Home
  • 523585 - Tris[(epoxypropoxypropyl)dimethylsilyloxy]-POSS®

EMAIL THIS PAGE TO A FRIEND
523585 Aldrich

Tris[(epoxypropoxypropyl)dimethylsilyloxy]-POSS®

Synonym: 3,7,14-Tris{[3-(epoxypropoxy)propyl]dimethylsilyloxy}-1,3,5,7,9,11,14-heptacyclopentyltricyclo[7.3.3.15,11]heptasiloxane

  • CAS Number 307496-38-4

  • Empirical Formula (Hill Notation) C59H114O18Si10

  • Molecular Weight 1392.38

  •  PubChem Substance ID 329757824

Purchase

Properties

Related Categories Incompletely Condensed, Materials Science, Nanomaterials, Silsesquioxanes: POSS Nanohybrids
InChI Key   SCSHXNSAPZJPEZ-UHFFFAOYSA-N
refractive index   n20/D >1.4850(lit.)

Description

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?