• USA Home
  • 525294 - Sucrose acetate isobutyrate solution

EMAIL THIS PAGE TO A FRIEND
525294 Aldrich

Sucrose acetate isobutyrate solution

90 wt. % in ethyl acetate

  • CAS Number 126-13-6

  • Empirical Formula (Hill Notation) C40H62O19

  • Molecular Weight 846.91

  •  MDL number MFCD00041859

  •  PubChem Substance ID 24874317

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
InChI Key   ZNEBZIJCDDCNRC-SWTLDUCYSA-N
concentration   90 wt. % in ethyl acetate
bp   78 °C
mp   −83 °C
density   1.11 g/mL at 25 °C

Description

Packaging

4 L in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1173 - class 3 - PG 2 - Ethyl acetate
WGK Germany 
1
Flash Point(F) 
69.8 °F
Flash Point(C) 
21 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?