• USA Home
  • 526231 - (R)-(−)-tert-Leucinol

EMAIL THIS PAGE TO A FRIEND
526231 Aldrich

(R)-(−)-tert-Leucinol

98%

Synonym: (R)-2-Amino-3,3-dimethyl-1-butanol

Purchase

Properties

Related Categories Amino Acids & Derivatives, Amino Alcohols, Chemical Biology, Chemical Synthesis, Peptide Chemistry More...
InChI Key   JBULSURVMXPBNA-YFKPBYRVSA-N
assay   98%
optical activity   [α]20/D −37°, c = 1.5 in ethanol
bp   70 °C/0.4 mmHg(lit.)
mp   30-33 °C(lit.)
density   0.9 g/mL at 25 °C(lit.)

Description

Packaging

100 mg in poly bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
194 °F
Flash Point(C) 
90 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?