EMAIL THIS PAGE TO A FRIEND
52639 Aldrich

Lutrol® OP 2000

non-ionic

Synonym: Polypropylene glycol 1450 oleate

Purchase

Description

Application

emollient, lubricant

Packaging

1 L in poly bottle

Legal Information

Lutrol is a registered trademark of BASF SE

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
>446 °F
Flash Point(C) 
>230 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?