• USA Home
  • 52720 - 1,4,7,10,13,16-Hexamethyl-1,4,7,10,13,16-hexaazacyclooctadecane

EMAIL THIS PAGE TO A FRIEND
52720 Aldrich

1,4,7,10,13,16-Hexamethyl-1,4,7,10,13,16-hexaazacyclooctadecane

≥97.0% (GC)

Synonym: Hexamethylhexacyclen

  • CAS Number 79676-97-4

  • Empirical Formula (Hill Notation) C18H42N6

  • Molecular Weight 342.57

  •  Beilstein Registry Number 4315531

  •  MDL number MFCD00075430

  •  PubChem Substance ID 57651379

Purchase

Properties

Related Categories Chelation/Complexation Compounds, Chemical Synthesis, Crown Ethers, Synthetic Reagents
InChI Key   KVXKXUJAIKWPHT-UHFFFAOYSA-N
assay   ≥97.0% (GC)
refractive index   n20/D 1.494(lit.)
  n20/D 1.494
bp   140 °C/0.02 mmHg(lit.)
density   0.944 g/mL at 25 °C(lit.)

Description

Other Notes

Hexacyclen-derivative forming stable sodides Cs+ and K+ (HMHCY)Na-. Efficient catalyst in reduction reactions

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?