• USA Home
  • 527572 - (E)-Phenyl-1,2-ethylenediboronic acid bis(pinacol) ester

EMAIL THIS PAGE TO A FRIEND
527572 Aldrich

(E)-Phenyl-1,2-ethylenediboronic acid bis(pinacol) ester

98%

Synonym: cis-1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl) styrene, cis-2-Phenylvinyl-1,2-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

Purchase

Properties

Related Categories Alkenyl Boronate Esters, Boronate Esters, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents More...
InChI Key   VQWACXAYGMFHOQ-PEZBUJJGSA-N
assay   98%
mp   69-73 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Arylboronic Acid-Pinacol Esters

The synthesis of biaryl compounds via the Suzuki coupling reaction has become more commonplace now that many arylboronic acids are readily available. Several years ago, Miyaura, et al. demonstrated t...
Chemfiles Volume 1 Article 1
Keywords: Coupling reactions, Suzuki coupling

Boronic Acid Esters

The synthesis of biaryl compounds via the Suzuki–Miyaura coupling reaction has become more commonplace now that many arylboronic acids are readily available. Several years ago, Miyaura et al. demonst...
Chemfiles Volume 4 Article 2
Keywords: Coupling reactions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?