• USA Home
  • 527645 - 2,3-Epoxy-2-methylbutane

EMAIL THIS PAGE TO A FRIEND
527645 Aldrich

2,3-Epoxy-2-methylbutane

97%

Synonym: 2-Methylbutylene-2-oxide

Purchase

Properties

Related Categories Epoxide Monomers, Materials Science, Monomers, Polymer Science
InChI Key   QPBYBLZYMNWGMO-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.3855(lit.)
bp   75 °C(lit.)
density   0.818 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BE 3 / PGII
WGK Germany 
2
RTECS 
EK3860000
Flash Point(F) 
-4 °F
Flash Point(C) 
-20 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?