• USA Home
  • 528927 - 1-Bromo-2,3,5-trichlorobenzene

EMAIL THIS PAGE TO A FRIEND
528927 Aldrich

1-Bromo-2,3,5-trichlorobenzene

96%

Purchase

Properties

Related Categories Aryl, Building Blocks, C6, Chemical Synthesis, Halogenated Hydrocarbons,
InChI Key   WOIGJFAVHOCDDW-UHFFFAOYSA-N
assay   96%
mp   58-61 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?