• USA Home
  • 529044 - 2,5-Dichlorobenzonitrile

EMAIL THIS PAGE TO A FRIEND
529044 Aldrich

2,5-Dichlorobenzonitrile

97%

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
InChI Key   LNGWRTKJZCBXGT-UHFFFAOYSA-N
assay   97%
mp   128-132 °C(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?