• USA Home
  • 52949 - 3-(2-Quinolyl)-DL-alanine

EMAIL THIS PAGE TO A FRIEND
52949 Aldrich

3-(2-Quinolyl)-DL-alanine

≥98.0% (HPLC)

  • CAS Number 123761-12-6

  • Empirical Formula (Hill Notation) C12H12N2O2

  • Molecular Weight 216.24

  •  Beilstein Registry Number 179632

  •  MDL number MFCD03095482

  •  PubChem Substance ID 57651391

Purchase

Properties

Related Categories Alanine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   CRSSRGSNAKKNNI-UHFFFAOYSA-N
assay   ≥98.0% (HPLC)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?