EMAIL THIS PAGE TO A FRIEND
530468 Aldrich

Acetonitrile-d3

99.8 atom % D

Synonym: Methyl-d3 cyanide, Trideuteroacetonitrile

  • CAS Number 2206-26-0

  • Linear Formula CD3CN

  • Molecular Weight 44.07

  •  Beilstein Registry Number 1740230

  •  MDL number MFCD00001881

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   WEVYAHXRMPXWCK-FIBGUPNXSA-N
isotopic purity   99.8 atom % D
contains   0.1% TMS
refractive index   n20/D 1.341(lit.)
bp   80.7 °C(lit.)
density   0.844 g/mL at 25 °C(lit.)
mass shift   M+3

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
42.8 °F
Flash Point(C) 
6 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

366544 Acetonitrile-d3, 99.8 atom % D, contains 0.03 % (v/v) TMS
613134 Acetonitrile-d3, 99.8 atom % D, contains 0.05 % (v/v) TMS

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?