• USA Home
  • 530514 - Trifluoroacetic acid-d

EMAIL THIS PAGE TO A FRIEND
530514 Aldrich

Trifluoroacetic acid-d

99.5 atom % D

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, NMR Solvents, Organic Acids,
InChI Key   DTQVDTLACAAQTR-DYCDLGHISA-N
isotopic purity   99.5 atom % D
mol wt   mol wt 115.02 by atom % calculation
packaging   pk of 10 × 0.75 mL
refractive index   n20/D 1.3(lit.)
bp   75 °C(lit.)
density   1.493 g/mL at 25 °C(lit.)
mass shift   M+1

Description

Packaging

1 pkg in ampule

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2699 8 / PGI
WGK Germany 
3
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?