• USA Home
  • 531898 - [(S,S)-Ethylenebis(4,5,6,7-tetrahydro-1-indenyl)]zirconium(IV)-(R)-1,1′-bi-2-naphtholate

EMAIL THIS PAGE TO A FRIEND
531898 Aldrich

[(S,S)-Ethylenebis(4,5,6,7-tetrahydro-1-indenyl)]zirconium(IV)-(R)-1,1′-bi-2-naphtholate

  • CAS Number 132881-66-4

  • Empirical Formula (Hill Notation) C40H36O2Zr

  • Molecular Weight 639.94

  •  MDL number MFCD03093919

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Other Chiral Catalysts
InChI Key   RENWIQQXMVJPLQ-UHFFFAOYSA-L
optical activity   [α]20/D +733°, c = 1 in methylene chloride
mp   157-162 °C(lit.)

Description

Packaging

100 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?