• USA Home
  • 532940 - (S)-(−)-1-[N-(1-Ethoxycarbonyl-3-phenylpropyl)-N-trifluoroacetyl]-L-lysine

EMAIL THIS PAGE TO A FRIEND
532940 Aldrich

(S)-(−)-1-[N-(1-Ethoxycarbonyl-3-phenylpropyl)-N-trifluoroacetyl]-L-lysine

97%

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Lysine Derivatives, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   QKGKAQLXEOARHX-HOTGVXAUSA-N
assay   97%
mp   133-138 °C(lit.)

Description

Packaging

1, 5 g in glass bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?