• USA Home
  • 533114 - N-Z-O-Benzyl-L-threonine

EMAIL THIS PAGE TO A FRIEND
533114 Aldrich

N-Z-O-Benzyl-L-threonine

97%

Synonym: (3R,2S)-3-Benzyloxy-2-benzyloxycarbonylaminobutyric acid

  • CAS Number 69863-36-1

  • Linear Formula CH3CH(OCH2C6H5)CH(NHCO2CH2C6H5)CO2H

  • Molecular Weight 343.37

  •  MDL number MFCD00077042

  •  PubChem Substance ID 24877942

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Threonine More...
InChI Key   FVKJXAYBJIAXAU-PBHICJAKSA-N
assay   97%
optical activity   [α]20/D −20°, c = 1 in H2O
mp   77-80 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?