• USA Home
  • 533149 - Nα-Boc-Nβ-xanthenyl-L-asparagine

EMAIL THIS PAGE TO A FRIEND
533149 Aldrich

Nα-Boc-Nβ-xanthenyl-L-asparagine

96%

Synonym: Nα-t-Boc-Nγ-Xanthyl-L-asparagine, Boc-Asn(Xan)-OH, Nα-Boc-Nγ-(9-xanthenyl)-L-asparagine

  • CAS Number 65420-40-8

  • Empirical Formula (Hill Notation) C22H24N2O6

  • Molecular Weight 412.44

  •  Beilstein Registry Number 5172403

  •  MDL number MFCD00066157

Purchase

Properties

Related Categories Asparagine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   YMGDQLXBNMRJMR-HNNXBMFYSA-N
assay   96%
optical activity   [α]20/D +3°, c = 1 in acetone
mp   177.5-181.5 °C(lit.)

Description

Packaging

1, 5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?