• USA Home
  • 533165 - Amberlite® IRC748I sodium form

EMAIL THIS PAGE TO A FRIEND
533165 Aldrich

Amberlite® IRC748I sodium form

Purchase

Properties

Related Categories Amberlite, Analytical/Chromatography, Cation, Chelating Ion Exchange Media, LPLC Separation Media,
description   weakly acidic cation exchange resin
limit   70 °C max. temp. (H+ form)
  90 °C max. temp. (Na+ form)
moisture   60-69%
matrix   styrene divinylbenzene (macroporous)
matrix active group   iminodiacetic acid functional group
particle size   23-30 mesh
  500-650 μm
operating pH   1.5 - 14
capacity   1.25 eq/L exchange capacity
  1.35 meq/mL(copper)

Description

Application

Chelating resin with high affinity for heavy metal cations over alkali or alkaline earth metals; ideal for use in non-aqueous media.

Useful in metal-chelation applications.

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Selection Guide for Polymeric Resins in Chromatography & Purification

Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?