• USA Home
  • 534889 - Amano Lipase PS-C II (immobilized on ceramic)

EMAIL THIS PAGE TO A FRIEND
534889 Aldrich

Amano Lipase PS-C II (immobilized on ceramic)

Purchase

Description

Application

One unit will produce 1.0 micromole of 1-Phenethyl alcohol to 1-Phenethyl acetate per min. at 25°C in the presence of vinyl acetate

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?