• USA Home
  • 534986 - (S)-(+)-(2-Butenoyl)-2,10-camphorsultam

EMAIL THIS PAGE TO A FRIEND
534986 Aldrich

(S)-(+)-(2-Butenoyl)-2,10-camphorsultam

97%

Synonym: (N-Crotonyl)-(2S)-bornane-10,2-sultam

  • CAS Number 94594-81-7

  • Empirical Formula (Hill Notation) C14H21NO3S

  • Molecular Weight 283.39

  •  MDL number MFCD00269665

  •  PubChem Substance ID 24878064

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Auxiliaries, Sulfur-Based
InChI Key   BKPQKSSKLBTRJO-LHNOPMQHSA-N
assay   97%
optical activity   [α]20/D +100°, c = 1 wt.% oxygen in ethanol
mp   182-185 °C(lit.)

Description

Packaging

500 mg in glass insert

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?