• USA Home
  • 535052 - 5-Methyl-1H-tetrazole

EMAIL THIS PAGE TO A FRIEND
535052 Aldrich

5-Methyl-1H-tetrazole

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Tetrazoles
InChI Key   XZGLNCKSNVGDNX-UHFFFAOYSA-N
assay   97%
mp   142-146 °C(lit.)

Description

Packaging

25 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
transport forbidden
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?