EMAIL THIS PAGE TO A FRIEND
537047 Aldrich

Propionic acid

99.5%

Synonym: Acid C3, Propanoic acid, Propanyl acid

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Cell Biology,
vapor density   2.55 (vs air)
vapor pressure   2.4 mmHg ( 20 °C)
InChI Key   XBDQKXXYIPTUBI-UHFFFAOYSA-N
assay   99.5%
autoignition temp.   955 °F
expl. lim.   12.1 %
refractive index   n20/D 1.386(lit.)
bp   141 °C(lit.)
mp   −24-−23 °C(lit.)
density   0.993 g/mL at 25 °C(lit.)

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN3463 - class 3 - Propionic acid
WGK Germany 
1
Flash Point(F) 
129.2 °F
Flash Point(C) 
54 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?