• USA Home
  • 537179 - (R)-(+)-1,1,1-Trifluoroheptan-2-ol

EMAIL THIS PAGE TO A FRIEND
537179 Aldrich

(R)-(+)-1,1,1-Trifluoroheptan-2-ol

97%

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   YFVHSPAONQXAIB-ZCFIWIBFSA-N
assay   97%
refractive index   n20/D 1.375(lit.)
bp   151-152 °C(lit.)
density   1.085 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1987C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
127.4 °F
Flash Point(C) 
53 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?