• USA Home
  • 537330 - Diethylamine, polymer-bound

EMAIL THIS PAGE TO A FRIEND
537330 Aldrich

Diethylamine, polymer-bound

100-200 mesh, extent of labeling: 1.0-1.5 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym: Benzyldiethylamine, polymer-bound, Diethylaminomethyl resin, Diethylaminomethyl-polystyrene

  • Linear Formula (C6H4CH2N(CH2CH3)2)x (CH2CH(C6H5))Y (C6H4(CHCH2)2)Z

  •  MDL number MFCD00144112

  •  PubChem Substance ID 24878212

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Organic Bases, Solid Supported Synthesis,
crosslinking   1 % cross-linked with divinylbenzene
extent of labeling   1.0-1.5 mmol/g loading
particle size   100-200 mesh

Description

Application

This tertiary amine base catalyzes acylation reactions. It also serves as a scavenger of protons and acids.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?