• USA Home
  • 537411 - N-(2,4-Dimethylphenyl)-3-oxobutyramide

EMAIL THIS PAGE TO A FRIEND
537411 Aldrich

N-(2,4-Dimethylphenyl)-3-oxobutyramide

99.5%

Synonym: AAMX, Acetoacet-m-xylidide

Purchase

Properties

Related Categories Amides, Building Blocks, C8 to C20+, Carbonyl Compounds, Chemical Synthesis,
InChI Key   HGVIAKXYAZRSEG-UHFFFAOYSA-N
assay   99.5%
mp   88-91 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
AK4585000
Flash Point(F) 
339.8 °F
Flash Point(C) 
171 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?