• USA Home
  • 538485 - 4-Nitrophenyl carbonate, polymer-bound on Wang Resin

EMAIL THIS PAGE TO A FRIEND
538485 Aldrich

4-Nitrophenyl carbonate, polymer-bound on Wang Resin

extent of labeling: 0.5-1.1 mmol/g loading, 100-200 mesh, 1 % cross-linked

Synonym: 4-Benzyloxybenzyl 4-nitrophenyl carbonate, polymer-bound, 4-Nitrophenyl carbonate, polymer-bound, Activated carbonate, polymer-bound

Purchase

Properties

Related Categories Chemical Synthesis, Other Resins, Solid Supported Synthesis, Specialty Synthesis
crosslinking   1 % cross-linked
extent of labeling   0.5-1.1 mmol/g loading
particle size   100-200 mesh

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?