• USA Home
  • 538590 - 3-Acetyl-6-bromocoumarin

EMAIL THIS PAGE TO A FRIEND
538590 Aldrich

3-Acetyl-6-bromocoumarin

97%

  • CAS Number 2199-93-1

  • Empirical Formula (Hill Notation) C11H7BrO3

  • Molecular Weight 267.08

  •  MDL number MFCD00024075

  •  PubChem Substance ID 24878344

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Coumarins, Halogenated Heterocycles, Heterocyclic Building Blocks More...
InChI Key   XFQYOFLFNKCHLG-UHFFFAOYSA-N
assay   97%
mp   225-229 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?