• USA Home
  • 538620 - Carboxyethylpolystryrene

EMAIL THIS PAGE TO A FRIEND
538620 Aldrich

Carboxyethylpolystryrene

100-200 mesh, extent of labeling: 0.8-1.5 mmol/g loading, 1 % cross-linked

Synonym: Carboxylated polystyrene

Purchase

Properties

Related Categories Chemical Synthesis, Other Resins, Solid Supported Synthesis, Specialty Synthesis
InChI Key   AWWLEBBAQLFDFA-UHFFFAOYSA-N
crosslinking   1 % cross-linked
extent of labeling   0.8-1.5 mmol/g loading
particle size   100-200 mesh

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?