• USA Home
  • 539805 - 2-Acetyl-4-nitro-1,3-indanedione

EMAIL THIS PAGE TO A FRIEND
539805 Aldrich

2-Acetyl-4-nitro-1,3-indanedione

97%

Synonym: 2-Acetyl-4-nitro-1H-indene-1,3(2H)-dione

  • CAS Number 25125-04-6

  • Empirical Formula (Hill Notation) C11H7NO5

  • Molecular Weight 233.18

  •  Beilstein Registry Number 2282627

  •  MDL number MFCD00728684

  •  PubChem Substance ID 24878424

Purchase

Properties

Related Categories Building Blocks, C11 to C12, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   OSZUSQXEYYPINX-UHFFFAOYSA-N
assay   97%
mp   145-148 °C(lit.)

Description

Application

2-Acetyl-4-nitro-1,3-indanedione may be used to synthesize 2-acetyl-4-hydroxyamino-1,3-indandione.

General description

2-Acetyl-4-nitro-1,3-indanedione can be prepared by employing 3-nitrophthaloyl chloride or 3-nitrophthalic anhydride as a starting reactant.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?