• USA Home
  • 541699 - 7-Acetyl-5,8-dihydroxy-4-isopropyl-1-methylbicyclo[4.3.0]nonane

EMAIL THIS PAGE TO A FRIEND
541699 Aldrich

7-Acetyl-5,8-dihydroxy-4-isopropyl-1-methylbicyclo[4.3.0]nonane

97%

  • CAS Number 457949-49-4

  • Empirical Formula (Hill Notation) C15H26O3

  • Molecular Weight 254.37

  •  PubChem Substance ID 24878550

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Ketones, Organic Building Blocks More...
InChI Key   JBUVDZNSKXFWKY-QMIVOQANSA-N
assay   97%
optical activity   [α]20/D +9°, c = 1 in chloroform
mp   48-50 °C(lit.)

Description

Packaging

100 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?