• USA Home
  • 541702 - 3-(4-Hydroxyphenyl)-N-benzylpropionamide

EMAIL THIS PAGE TO A FRIEND
541702 Aldrich

3-(4-Hydroxyphenyl)-N-benzylpropionamide

96%

Purchase

Properties

Related Categories Amides, Building Blocks, C8 to C20+, Carbonyl Compounds, Chemical Synthesis,
InChI Key   OSPJHCUBEDDXIQ-UHFFFAOYSA-N
assay   96%
mp   111-114 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?