• USA Home
  • 541885 - 1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane

EMAIL THIS PAGE TO A FRIEND
541885 Aldrich

1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane

97%

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   XYMBGGTVDWPIBA-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.4(lit.)
bp   152-153 °C(lit.)
density   1.991 g/mL at 25 °C(lit.)

Description

Packaging

1, 5 g in glass bottle

General description

1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane is a halogenated hydrocarbon.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?