• USA Home
  • 543411 - 2-Chlorobenzhydrol, polymer-bound

EMAIL THIS PAGE TO A FRIEND
543411 Aldrich

2-Chlorobenzhydrol, polymer-bound

100-200 mesh, extent of labeling: 1.0-2.0 mmol/g loading, 1 % cross-linked

Synonym: 2-Chlorodiphenylmethanol resin, Hydroxy-(2-chlorophenyl)methyl polystyrene

Purchase

Properties

Related Categories Chemical Synthesis, Other Resins, Solid Supported Synthesis, Specialty Synthesis
crosslinking   1 % cross-linked
extent of labeling   1.0-2.0 mmol/g loading
particle size   100-200 mesh

Description

Packaging

1, 25 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?