CAS Number 4348-68-9
Empirical Formula (Hill Notation) C26H21BrO7
Molecular Weight 525.34
Beilstein Registry Number 98941
MDL number MFCD03791085
PubChem Substance ID 24878701
| Related Categories | Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis |
| assay | ≥96.0% (HPLC) |
| storage temp. | 2-8°C |
Building block for the synthesis of unnatural arabinofuranosyl nucleosides1,2
5 g in poly bottle
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?