• USA Home
  • 543713 - 6-Fluoro-4-methylcoumarin-3-carbonitrile

EMAIL THIS PAGE TO A FRIEND
543713 Aldrich

6-Fluoro-4-methylcoumarin-3-carbonitrile

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Coumarins, Heterocyclic Building Blocks
InChI Key   PVZRETKTYNNESC-UHFFFAOYSA-N
assay   97%
mp   164-168 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?