• USA Home
  • 543829 - 6-Fluoro-8-nitrochromone-3-carboxaldehyde

EMAIL THIS PAGE TO A FRIEND
543829 Aldrich

6-Fluoro-8-nitrochromone-3-carboxaldehyde

97%

Purchase

Properties

Related Categories Aldehydes, Benzopyrans, Building Blocks, C10-C12, Carbonyl Compounds,
InChI Key   MFTIKIFELZBYLH-UHFFFAOYSA-N
assay   97%
mp   110-114 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?