• USA Home
  • 544035 - 2-Pyridyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside

EMAIL THIS PAGE TO A FRIEND
544035 Aldrich

2-Pyridyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside

97%

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
InChI Key   MGGZRMLMASJEHA-FTWQHDNSSA-N
assay   97%
optical activity   [α]20/D −28°, c = 1 in chloroform
mp   127-131 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?