• USA Home
  • 545252 - S-(2-Phenoxyethyl)thioacetate

EMAIL THIS PAGE TO A FRIEND
545252 Aldrich

S-(2-Phenoxyethyl)thioacetate

98%

Synonym: Thioacetic acid, (S)-(2-phenoxy-ethyl ester)

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiol Esters More...
InChI Key   MDZLAKVKAKOCPS-UHFFFAOYSA-N
assay   98%
mp   47-51 °C(lit.)

Description

Packaging

1 g in glass bottle

General description

S-(2-Phenoxyethyl)thioacetate, also known as S-(2-phenoxyethyl) ethanethioate, can be prepared from thiirane and phenyl acetate.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?