• USA Home
  • 546690 - 2,4,5-Trifluoro-3-methoxybenzoyl chloride

EMAIL THIS PAGE TO A FRIEND
546690 Aldrich

2,4,5-Trifluoro-3-methoxybenzoyl chloride

97%

Purchase

Properties

Related Categories Aldehydes, Aryl Fluorinated Building Blocks, Building Blocks, C7-C8, C8,
InChI Key   JVQSZTJTWSUJCR-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.503(lit.)
density   1.472 g/mL at 25 °C(lit.)

Description

Packaging

25 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1760 8 / PGII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?