• USA Home
  • 547379 - StratoSpheres PL-AMS resin

EMAIL THIS PAGE TO A FRIEND
547379 Aldrich

StratoSpheres PL-AMS resin

100-200 mesh, extent of labeling: ~1.0 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym: (Aminomethyl)polystyrene, AM-polystyrene, Benzylamine polymer-bound, Poly(styrene-co-divinylbenzene), aminomethylated, Poly(styrene-co-vinylbenzylamine-co-divinylbenzene, Polystyrene crosslinked with divinylbenzene, aminomethylated

Purchase

Properties

Related Categories Chemical Synthesis, Solid Supported Synthesis, Specialty Synthesis, StratoSpheres
crosslinking   1 % cross-linked with divinylbenzene
extent of labeling   ~1.0 mmol/g loading
particle size   100-200 mesh
storage temp.   2-8°C

Description

Application

Scavenges acids, acid chlorides, anhydrides, aldehydes and other electrophiles

Legal Information

StratoSpheres is a trademark of Agilent Technologies, Inc.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?