• USA Home
  • 547506 - StratoSpheres PL-FMP resin

EMAIL THIS PAGE TO A FRIEND
547506 Aldrich

StratoSpheres PL-FMP resin

50-100 mesh, extent of labeling: 1.6 mmol/g loading, 1 % cross-linked

Synonym: (4-Formyl-3-methoxyphenoxymethyl)polystyrene, 4-Formyl-3-methoxyphenoxy resin, 4-Hydroxy-2-methoxybenzaldehyde, polymer-bound, AMEBA resin, Aldehyde resin

Purchase

Properties

Related Categories Chemical Synthesis, Solid Supported Synthesis, Specialty Synthesis, StratoSpheres
crosslinking   1 % cross-linked
extent of labeling   1.6 mmol/g loading
particle size   50-100 mesh

Description

Packaging

5, 25 g in glass bottle

Legal Information

StratoSpheres is a trademark of Agilent Technologies, Inc.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?