Linear Formula [[(CH3)3Si]2N]3Yb
Molecular Weight 654.19
MDL number MFCD03427095
PubChem Substance ID 24878987
| Related Categories | CVD and ALD Precursors by Metal, Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Micro/NanoElectronics, |
| assay | 98% |
| form | solid |
| mp | 162-164 °C(lit.) |
1 g in glass bottle
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?