• USA Home
  • 548901 - 3-(6-Methyl-2H-1,4-benzoxazin-3(4H)-one-4-yl)-propionitrile

EMAIL THIS PAGE TO A FRIEND
548901 Aldrich

3-(6-Methyl-2H-1,4-benzoxazin-3(4H)-one-4-yl)-propionitrile

97%

Purchase

Properties

Related Categories Benzoxazines, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
InChI Key   SZEGFLHUNYLAOJ-UHFFFAOYSA-N
assay   97%
mp   128-132 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?