• USA Home
  • 550833 - 2-(N-Phenyl-N-ethyl)aminoethylpyridinium chloride

EMAIL THIS PAGE TO A FRIEND
550833 Aldrich

2-(N-Phenyl-N-ethyl)aminoethylpyridinium chloride

97%

  • CAS Number 14408-19-6

  • Empirical Formula (Hill Notation) C15H19ClN2

  • Molecular Weight 262.78

  •  MDL number MFCD00036054

  •  PubChem Substance ID 24879074

Purchase

Properties

Related Categories Building Blocks, C13 to C15, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   BKROMFFRLPSNBH-UHFFFAOYSA-M
assay   97%
mp   82-86 °C(lit.)

Description

Packaging

25 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?