• USA Home
  • 551058 - (S)-4,5,5-Triphenyl-2-oxazolidinone

EMAIL THIS PAGE TO A FRIEND
551058 Aldrich

(S)-4,5,5-Triphenyl-2-oxazolidinone

97%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Auxiliaries, Oxazolidinone Derivatives
InChI Key   QCWBJFYOEHCELV-IBGZPJMESA-N
assay   97%
optical activity   [α]20/D −216°, c = 1 in chloroform
mp   245-249 °C(lit.)

Description

Packaging

1, 5 g in glass bottle

Legal Information

A Product of Onyx Scientific, U.K.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Oxazolidinone Chiral Auxiliaries in Chemical Sythesis

Oxazolidinone chiral auxiliaries are often used for diastereoselective Michael additions, alkylations, aldol condensations, cyclopropanations, Diels-Alder, and other reactions. These new offerings co...
Keywords: Acylations, Addition reactions, Alkylations, Applications, Catalog, Chiral auxiliaries, Condensations, Cyclopropanations, Filtration, Type

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?