-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
Synonym:
N-
CAS Number 852527-48-1
Empirical Formula (Hill Notation) C13H9F17INO
Molecular Weight 645.09
MDL number MFCD09265203
PubChem Substance ID 24879285
| Related Categories | Chemical Synthesis, Fluorous Synthesis, Rf-Tagged Building Blocks and Precursors, Specialty Synthesis |
| assay | ≥97.0% |
5, 25 mg in glass btl
Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?