• USA Home
  • 553042 - α-(Trimethylsilyloxy)phenylacetonitrile

EMAIL THIS PAGE TO A FRIEND
553042 Aldrich

α-(Trimethylsilyloxy)phenylacetonitrile

97%

Synonym: Phenyl-trimethylsilanyloxy-acetonitrile

Purchase

Properties

Related Categories Building Blocks, C10 to C27, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
InChI Key   DTAFQWDNWAXRLX-UHFFFAOYSA-N
assay   97%
bp   91 °C/0.5 mmHg(lit.)
density   0.978 g/mL at 25 °C(lit.)

Description

Packaging

1 mL in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?