• USA Home
  • 554618 - (R)-(+)-2-Amino-4-methyl-1,1-diphenylpentane

EMAIL THIS PAGE TO A FRIEND
554618 Aldrich

(R)-(+)-2-Amino-4-methyl-1,1-diphenylpentane

98%

Purchase

Properties

Related Categories Amines, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   ICJINWKVCCYYQW-QGZVFWFLSA-N
assay   98%
optical activity   [α]20/D +35°, c = 1 in chloroform
mp   50-53 °C(lit.)

Description

Packaging

500 mg in glass bottle

Legal Information

A Product of Onyx Scientific, U.K.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?