• USA Home
  • 554820 - StratoSpheres PL-DIPAM (Diisopropylaminomethyl) resin

EMAIL THIS PAGE TO A FRIEND
554820 Aldrich

StratoSpheres PL-DIPAM (Diisopropylaminomethyl) resin

50-100 mesh, extent of labeling: 3.0 mmol/g loading, 1 % cross-linked

Synonym: ‘Hünig’s base’ on polystyrene, Benzyldiisopropylamine, polymer-bound, Copolymer of styrene and divinylbenzene, diisopropylaminomethylated, Diisopropylamine, polymer-bound, Diisopropylaminomethyl-polystyrene, Polystyrene cross-linked with divinylbenzene, diisopropylaminomethylated

Purchase

Properties

Related Categories Chemical Synthesis, Solid Supported Synthesis, Specialty Synthesis, StratoSpheres
crosslinking   1 % cross-linked
extent of labeling   3.0 mmol/g loading
particle size   50-100 mesh

Description

Application

Used in conjunction with polymer-bound isocyanates to scavenge amines.

Packaging

5, 25 g in glass bottle

Legal Information

StratoSpheres is a trademark of Agilent Technologies, Inc.

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?