• USA Home
  • 554901 - 1,4-Di(octyloxy)benzene

EMAIL THIS PAGE TO A FRIEND
554901 Aldrich

1,4-Di(octyloxy)benzene

98%

Purchase

Properties

Related Categories Building Blocks, C22 to C40+, Chemical Synthesis, Ethers, Organic Building Blocks,
InChI Key   KFQGWEDSKAPIDJ-UHFFFAOYSA-N
assay   98%
mp   56-59 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?